missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II Antibody (HIS19), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-00462JF646
This item is not returnable.
View return policy
Description
MHC Class II Monoclonal specifically detects MHC Class II in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| MHCClassII | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HLA-DMA | |
| The immunogen for this antibody is MHC Class II. | |
| 0.1 mL | |
| Primary | |
| Rat | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| HIS19 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
| Mouse | |
| Protein A or G purified | |
| Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Major Histocompatibility Complexes, Transcription Factors and Regulators | |
| 3108 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction