missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II Antibody (P-TH81A5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61033
This item is not returnable.
View return policy
Description
MHC Class II Monoclonal specifically detects MHC Class II in Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| MHC Class II | |
| Monoclonal | |
| 1 mg/mL | |
| Flow Cytometry 1:10 - 1:1000 | |
| class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
| Mouse | |
| Unpurified | |
| Adaptive Immunity, Asthma, B Cell Development and Differentiation Markers, Cell Biology, Diabetes Research, Immune System Diseases, Immunology, Major Histocompatibility Complexes, Transcription Factors and Regulators | |
| 3108 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Flow Cytometry, Immunohistochemistry, Functional Assay | |
| P-TH81A5 | |
| Unconjugated | |
| Ascites with 0.09% Sodium Azide | |
| HLA-DMA | |
| Thymoctes from: cattle, horse, goat, rat | |
| 0.1 mg | |
| Primary | |
| Pig |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?