Learn More
MHC Class II (I-A/I-E) Rat anti-Mouse, APC, Clone: M5/114.15.2, Proteintech
Rat Monoclonal Antibody
Brand:  Proteintech APC-65122-100UG
Product Code. 16820506
  • € 114.00 / 100µg
Description
The MHC Class II (I-A/I-E) antibody from Proteintech is a rat monoclonal antibody generated with activated c57bl/6 mouse spleen cells of mouse MHC Class II (I-A/I-E). This antibody recognizes mouse antigen. The MHC Class II (I-A/I-E) antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
Specifications
MHC Class II (I-A/I-E)
Monoclonal
0.2 mg/mL
Rat
Primary
Mouse
Flow Cytometry
M5/114.15.2
APC
100 μg
14960
IgG2b