missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MHC Class II RT1B Antibody (OX-6), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-65541DL594
This item is not returnable.
View return policy
Description
MHC Class II RT1B Monoclonal antibody specifically detects MHC Class II RT1B in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry Free-Floating, CyTOF
Specifications
| MHC Class II RT1B | |
| Monoclonal | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| OX-6 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, CyTOF-ready | |
| DMA, DMB, DP beta 1, DPB 1, DPB1, DRB, HLA class II histocompatibility antigen DM beta chain, HLA DMB, HLA DPB1, HLADM, HLADP1B, Major histocompatibility complex class II, Major histocompatibility complex class II DP beta 1, MHC class II antigen DMB, MHC DPB1, RING6, RING7 | |
| Rat thymocyte membrane glycoproteins | |
| 0.25 mL | |
| Adaptive Immunity, Immunology | |
| 24738 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction