missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MHC I Monomorphic Monoclonal Antibody (CVS22), PE
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA528476
This item is not returnable.
View return policy
Description
Mouse anti Horse MHC Class I Monomorphic antibody, clone CVS22 recognizes monomorphic equine MHC Class I and was classified at the International Equine Leucocyte Antigen Workshop. Reconstitute by adding 1 mL distilled water.
MHC I Monomorphic is part of major histocompatibility complex (MHC) is a cluster of genes that are important in the immune response to infections. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
Specifications
| MHC I Monomorphic | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 5% sucrose and 0.09% sodium azide | |
| 0 | |
| EQMHCB2, EQMHCC1, MHCB1, MHCX1 | |
| Equine leucocytes. | |
| 100 Tests | |
| Primary | |
| Horse | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| CVS22 | |
| PE | |
| MHCX1 | |
| 3349.1; BOLA; BoLA-N; BoLA-NC; Bota; classical MHC class I antigen; D18.5; Eqca-1; EQMHCB2; EQMHCC1; HD15; hypothetical protein LOC509597; LA-A11; major histocompatibility complex; major histocompatibility complex, class I, A; MHC class 1; MHC class I antigen; MHC class I antigen 10.b; MHC class I antigen 2*02501; MHC class I antigen 2.5; MHC class I antigen 3.1; MHC class I antigen 3.5; MHC class I antigen 3.6; MHC class I antigen 9.5; MHC class I antigen 9.6; MHC class I antigen clone 2; MHC class I antigen heavy chain; MHC class I heavy chain; MHC class I heavy chain clone 302D.1m; MHCB1; MHCX1; non-classical MHC class I antigen; non-MHC class I antigen; uncharacterized protein LOC790811 | |
| Mouse | |
| Protein A | |
| RUO | |
| 100034209, 100034210, 100034211, 100056391 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction