missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MI-136, MedChemExpress
Description
MI-136 is an inhibitor of the menin-MLL protein-protein interaction (PPI), with an IC50 of 31 nM and a Kd of 23.6 nM. MI-136 shows to block AR signaling and has the potential for the study in castration-resistant tumors.
Specifications
Specifications
| Chemical Name or Material | MI-136 |
| CAS | 1628316-74-4 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H21F3N6S |
| Quantity | 100 mg |
| Solubility Information | DMSO : 110 mg/mL (233.79 mM; Need ultrasonic) |
| SMILES | FC(F)(F)CC(S1)=CC2=C1N=CN=C2NC3CCN(CC4=CC=C(NC(C#N)=C5)C5=C4)CC3 |
| Molecular Weight (g/mol) | 470.51 |
| Formula Weight | 470.51 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?