missing translation for 'onlineSavingsMsg'
Learn More

MI-136, MedChemExpress

Product Code. 16430188
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16430188 5 mg 5mg
16467586 100 mg 100mg
16453697 50 mg 50mg
16407728 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16430188 Supplier Medchem Express Supplier No. HY19319

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MI-136 is an inhibitor of the menin-MLL protein-protein interaction (PPI), with an IC50 of 31 nM and a Kd of 23.6 nM. MI-136 shows to block AR signaling and has the potential for the study in castration-resistant tumors.

MI-136 is an inhibitor of the menin-MLL protein-protein interaction (PPI), with an IC50 of 31 nM and a Kd of 23.6 nM. MI-136 shows to block AR signaling and has the potential for the study in castration-resistant tumors.

Tekniske data

Chemical Name or Material MI-136
CAS 1628316-74-4
Color White
Purity Grade Notes Research
Molecular Formula C23H21F3N6S
Quantity 5 mg
Solubility Information DMSO : 110 mg/mL (233.79 mM; Need ultrasonic)
SMILES FC(F)(F)CC(S1)=CC2=C1N=CN=C2NC3CCN(CC4=CC=C(NC(C#N)=C5)C5=C4)CC3
Molecular Weight (g/mol) 470.51
Formula Weight 470.51
Percent Purity 99.71%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.