missing translation for 'onlineSavingsMsg'
Learn More

MI-3, MedChemExpress

Product Code. 16436676
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16436676

Brand: Medchem Express HY15223

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MI-3 (Menin-MLL inhibitor 3) is a potent and high affinity menin-MLL inhibitor with an IC50 of 648 nM and a Kd of 201 nM.

MI-3 (Menin-MLL inhibitor 3) is a potent and high affinity menin-MLL inhibitor with an IC50 of 648 nM and a Kd of 201 nM.

Specifications

Chemical Name or Material MI-3
Physical Form Solid
CAS 1271738-59-0
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H25N5S2
For Use With (Application) Cancer-programmed cell death
Synonym Menin-MLL inhibitor 3
Solubility Information DMSO : 8.33 mg/mL (22.18 mM; Need ultrasonic)
SMILES CC(C)C1=CC2=C(N3CCN(C4=NCC(C)(C)S4)CC3)N=CN=C2S1
Molecular Weight (g/mol) 375.55
Formula Weight 375.55
Percent Purity 99.51%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.