missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MIA Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559957
This item is not returnable.
View return policy
Description
Immunogen sequence: PKLADRKLCA DQECSHPISM AVALQDYMAP DCRFLTIHRG QVVYVFSKLK GRGRLFWGGS VQGDYYGDLA ARLGYFPSSI VREDQTLKPG KVDVKTDKWD FYC Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.
Tumorigenesis is a process that is mediated by a network of membrane, cytosolic and nuclear associated factors, which regulate proliferation and cell-matrix interaction through signaling cascades. The phenotype of malignant melanomas in vivo depends on the global expression of stimulatory or inhibitory factors generated in both the tumors cells and their environment. One example, Melanoma inhibitory activity (cartilage-derived retinoic acid-sensitive protein (CD-RAP), MIA) is a Src homology 3 (SH3)-like domain containing protein that is secreted from chondrocytes and malignant melanoma cells. MIA is translated as a 131-amino acid pro-form and processed into a mature 107-amino acid protein after cleavage of a secretion signal. MIA is expressed during chondrogenesis and in mature chondrocytes, suggesting that MIA is necessary for normal cartilage cell phenotype. MIA mRNA is present in carcinomas of the colon, ovary, kidney, and head/neck, and may represent a marker to monitor melanomic activity.
Specifications
| MIA | |
| Polyclonal | |
| Unconjugated | |
| MIA | |
| cartilage derived retinoic acid sensitive protein; cartilage-derived retinoic acid-sensitive protein; Cdrap; CD-RAP; melanoma inhibitory activity; melanoma inhibitory activity 1; melanoma inhibitory activity protein; melanoma inhibitory activity/condrocyte-derived retinoic acid sensitive protein homolog (MIA/CD-RAP); melanoma-derived growth regulatory protein; melanoma-inhibitory-activity; MIA; Mia1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 8190 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q16674 | |
| MIA | |
| Recombinant protein corresponding to Human MIA. Recombinant protein control fragment (Product #RP-90572). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction