missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ MIA3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29497
This item is not returnable.
View return policy
Description
MIA3 Polyclonal antibody specifically detects MIA3 in Human samples. It is validated for Western Blot
Specifications
| MIA3 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| ARNT, C219 Reactive Peptide, C219-Reactive Peptide, D320, KIAA0268, Melanoma Inhibitory Activity Family, Member 3, Melanoma Inhibitory Activity Protein 3, TANGO, TANGO1, Transport And Golgi Organization, Transport And Golgi Organization Protein 1, UNQ6077 | |
| Between 350 and 400 | |
| 100 μg | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 375056 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction