Learn More
Mibefradil dihydrochloride, MedChemExpress
Product Code. 16492707
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16492707
Brand: Medchem Express HY15553A
MedChemExpress Mibefradil dihydrochloride (Ro 40-5967 dihydrochloride) is a calcium channel blocker with moderate selectivity for T-type Ca2+ channels (IC50s of 2.7 μM and 18.6 μM for T-type and L-type currents, respectively).
Mibefradil dihydrochloride (Ro 40-5967 dihydrochloride) is a calcium channel blocker with moderate selectivity for T-type Ca2+ channels (IC50s of 2.7 μM and 18.6 μM for T-type and L-type currents, respectively).
Specifications
Chemical Name or Material Mibefradil dihydrochloride
CAS 116666-63-8
Color White
Purity Grade Notes Research
Molecular Formula C29H40Cl2FN3O3
Quantity 50 mg
Synonym Ro 40-5967 dihydrochloride
Solubility Information H2O : ≥ 125 mg/mL (219.86 mM)
SMILES O=C(O[C@@]1(CCN(CCCC2=NC3=CC=CC=C3N2)C)[C@@H](C(C)C)C4=C(C=C(F)C=C4)CC1)COC.[H]Cl.[H]Cl
Molecular Weight (g/mol) 568.55
Formula Weight 568.55
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only