missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MIC-1 Polyclonal Antibody

MIC-1 Polyclonal Antibody
Brand: Invitrogen PA5142271
This item is not returnable.
View return policy
Description
MIC-1 Polyclonal Antibody for Western Blot, IHC (P)
Specifications
| MIC-1 | |
| Polyclonal | |
| Unconjugated | |
| Rmc1 | |
| 2400010D15Rik; 3110002H16Rik; C18orf8; chromosome 18 open reading frame 8; colon cancer-associated protein Mic1; colon cancer-associated protein Mic1 homolog; HsT2591; macrophage inhibitory cytokine 1; Mic1; Mic-1; Regulator of MON1-CCZ1 complex; RGD1311805; RIKEN cDNA 3110002H16 gene; RMC1; similar to RIKEN cDNA 2400010D15; uncharacterized protein C18orf8; uncharacterized protein C18orf8 homolog; WD repeat-containing protein 98; WDR98 | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 29919 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q96DM3 | |
| Rmc1 | |
| Peptide with sequence C-DKLNHEYKKYLDAEQ. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction