missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mic1 Antibody (OTI4E4), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72668R
This item is not returnable.
View return policy
Description
Mic1 Monoclonal specifically detects Mic1 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| Mic1 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RMC1 | |
| Full length recombinant protein of human Mic1 (NP_037458) produced in HEK293T cells. | |
| 0.1 mL | |
| 29919.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI4E4 | |
| Western Blot, Flow Cytometry | |
| chromosome 18 open reading frame 8, Colon cancer-associated protein Mic1, FLJ23453, HsT2591, Mic-1, MIC1colon cancer associated protein Mic1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction