missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mic1 Antibody (OTI4E4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-01949
This item is not returnable.
View return policy
Description
Mic1 Monoclonal specifically detects Mic1 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| Mic1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000, Flow Cytometry 1:100 | |
| RMC1 | |
| Full length human recombinant protein of human C18orf8(NP_037458) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| 29919 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Flow Cytometry | |
| OTI4E4 | |
| Unconjugated | |
| chromosome 18 open reading frame 8, Colon cancer-associated protein Mic1, FLJ23453, HsT2591, Mic-1, MIC1colon cancer associated protein Mic1 | |
| Mouse | |
| 74.8 kDa | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction