missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICA Antibody (159227), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1300MFV500
This item is not returnable.
View return policy
Description
MICA Monoclonal antibody specifically detects MICA in Human samples. It is validated for Western Blot, Flow Cytometry, Blocking Assay, CyTOF, KnockDown
Specifications
| MICA | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 100507436 | |
| Store at 4C in the dark. |
| Western Blot, Flow Cytometry, Blocking Assay, CyTOF, KnockDown | |
| 159227 | |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready, Knockout Validated | |
| FLJ60820, MGC111087, PERB11.1 | |
| BaF3 mouse pro-B cell line transfected with human MICA | |
| RUO | |
| Primary | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction