Learn More
MICA/B Antibody (159207) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB13001JF525
Product Code. 30176892
  • ONLINE EXCLUSIVE PRICE
    € 650.00
    € 614.25 / 0.10mL
    Save € 35.75
    5% Off
Description
MICA/B Monoclonal antibody specifically detects MICA/B in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Blockade of Receptor-ligand Interaction,CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MICA/B
Monoclonal
Janelia Fluor 525
MHC class I chain-related protein A, MHC class I chain-related protein B, MIC, MICA, MICB, PERB11, PERB11.1, PERB11.2
BaF3 mouse pro-B cell line transfected with human MICA, Accession # Q29983
RUO
100507436
Store at 4°C in the dark.
IgG2a
Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF
159207
50mM Sodium Borate
Mouse
0.1 mL