missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICA/B Antibody (159207) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB13001JF635
This item is not returnable.
View return policy
Description
MICA/B Monoclonal antibody specifically detects MICA/B in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Blockade of Receptor-ligand Interaction,CyTOF
Specifications
| MICA/B | |
| Monoclonal | |
| Janelia Fluor 635 | |
| MHC class I chain-related protein A, MHC class I chain-related protein B, MIC, MICA, MICB, PERB11, PERB11.1, PERB11.2 | |
| BaF3 mouse pro-B cell line transfected with human MICA, Accession # Q29983 | |
| RUO | |
| 100507436 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF | |
| 159207 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction