Learn More
MICA/B Antibody (159207), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB13001MFV500
Product Code. 18519266
  • € 684.00 / 0.10mL
Description
MICA/B Monoclonal antibody specifically detects MICA/B in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Blocking Assay, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MICA/B
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
0.1 mL
Primary
Human
IgG2a
Flow Cytometry, Immunohistochemistry, Blocking Assay, CyTOF
159207
Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, CyTOF-ready
MHC class I chain-related protein A, MHC class I chain-related protein B, MIC, MICA, MICB, PERB11, PERB11.1, PERB11.2
BaF3 mouse pro-B cell line transfected with human MICA, Accession # Q29983
RUO
100507436
Store at 4C in the dark.