Learn More
MICA/B Antibody (159207), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB13001PECY7
This item is not returnable.
View return policy
Description
MICA/B Monoclonal antibody specifically detects MICA/B in Human samples. It is validated for Flow Cytometry
Specifications
| MICA/B | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 100507436 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG2a |
| Flow Cytometry | |
| 159207 | |
| Flow Cytometry | |
| MHC class I chain-related protein A, MHC class I chain-related protein B, MIC, MICA, MICB, PERB11, PERB11.1, PERB11.2 | |
| BaF3 mouse pro-B cell line transfected with human MICA, Accession # Q29983 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |