missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MICA/B Polyclonal Antibody
GREENER_CHOICE

Product Code. 17177253
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17177253 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17177253 Supplier Invitrogen™ Supplier No. PA5109323

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: U937, Mouse bone marrow, A431, rat skin tissue, human lung cancer tissue, human colon tissue, human breast carcinoma tissue, mouse stomach tissue.

MICA encodes the higly polymorphic MHC (HLA) class I chain-related gene A. The protein product is expressed on the cell surface, although unlike canonical class I molecules does not seem to associate with beta-2-microglobulin. It is thought that MICA functions as a stress-induced antigen that is broadly recognized by intestinal epithelial gamma delta T cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MICA/B
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene MICA
Gene Accession No. Q29980, Q29983
Gene Alias HLA class I antigen; MHC class I chain-related protein A; MHC class I chain-related protein B; MHC class I mic-B antigen; MHC class I polypeptide-related sequence A; MHC class I polypeptide-related sequence B; MHC class I related sequence A; MHC class I-like molecule PERB11.2-IMX; MICA; MIC-A; MICB; MIC-B; PERB11.1; PERB11.2; sMICA; sMICB; soluble MICA; soluble MICB; stress inducible class I homolog; truncated MHC class I polypeptide-related sequence A
Gene Symbols MICA, MICB
Host Species Rabbit
Immunogen Recombinant protein within Human MICA/MICB aa 182-325.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100507436, 4277
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.