missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICAL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-85141-25ul
This item is not returnable.
View return policy
Description
MICAL1 Polyclonal antibody specifically detects MICAL1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| MICAL1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/mL, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| CasL interacting molecule, DKFZp434B1517, FLJ11937, FLJ21739, MICALMolecule interacting with CasL protein 1, microtubule associated monoxygenase, calponin and LIM domain containing 1, NEDD9 interacting protein with calponin homology and LIM domains, NEDD9-interacting protein with calponin homology and LIM domains, NICALMICAL-1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: VRDLYDVLAKEPVQRNNDKTDTGMPATGSAGTQEELLRWCQEQTAGYPGVHVSDLSSSWADGLALCALVYRLQPGLLEPSELQGLGALEATAWALKV | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 64780 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction