missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICAL1 Antibody (OTI6H4), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72670MFV500
This item is not returnable.
View return policy
Description
MICAL1 Monoclonal antibody specifically detects MICAL1 in Human samples. It is validated for Flow Cytometry
Specifications
| MICAL1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 64780 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry | |
| OTI6H4 | |
| Flow Cytometry | |
| CasL interacting molecule, DKFZp434B1517, FLJ11937, FLJ21739, MICALMolecule interacting with CasL protein 1, microtubule associated monoxygenase, calponin and LIM domain containing 1, NEDD9 interacting protein with calponin homology and LIM domains, NEDD9-interacting protein with calponin homology and LIM domains, NICALMICAL-1 | |
| Full length human recombinant protein of human MICAL1 (NP_073602) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction