Learn More
MICAL1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech CL488-14818-20UL
Description
The MICAL1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MICAL1. This antibody recognizes human, mouse antigen. The MICAL1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| MICAL1 | |
| 1000 μg/mL | |
| DKFZp434B1517, FLJ11937, FLJ21739, MICAL, MICAL 1, MICAL1, NICAL | |
| 20 μL | |
| 64780 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence | |
| CoraLite Plus 488 | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |