Learn More
MICAL1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech CL488-14818-20UL
 View more versions of this product
Product Code. 18090326
  • € 145.00 / 20µL
Description
The MICAL1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MICAL1. This antibody recognizes human, mouse antigen. The MICAL1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
MICAL1
1000 μg/mL
DKFZp434B1517, FLJ11937, FLJ21739, MICAL, MICAL 1, MICAL1, NICAL
20 μL
64780
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Liquid
Immunofluorescence
CoraLite Plus 488
Rabbit
Primary
Human, Mouse
150 kDa