missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICAL2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 409.50 - € 646.80
Specifications
| Antigen | MICAL2 |
|---|---|
| Dilution | Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18458420
|
Novus Biologicals
NBP1-81483-25ul |
25ul |
€ 433.00 € 409.50 / 25µL Save € 23.50 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18278115
|
Novus Biologicals
NBP1-81483 |
0.1 mL |
€ 684.00 € 646.80 / 0.10mL Save € 37.20 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MICAL2 Polyclonal specifically detects MICAL2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MICAL2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 9645 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PVTTGKEMASAQEPDKLSMVMYLSKFYELFRGTPLRPVDSWRKNYGENADLSLAKSSISNNYLNLTFPRKRTPRVDGQTGENDMNKRRRKGFTNLDEPSNFSSRSLGSNQECGSSKEGGNQNKVKSM | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| DKFZp686H03148, DKFZp686H2469, flavoprotein oxidoreductase MICAL2, KIAA0750MICAL2PV2, MICAL2PV1, microtubule associated monoxygenase, calponin and LIM domain containing 2, protein MICAL-2 | |
| MICAL2 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title