missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MICALL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524826
This item is not returnable.
View return policy
Description
MICALL2 Polyclonal Antibody for Western Blot, IHC (P), Flow
Effector of small Rab GTPases which is involved in junctional complexes assembly through the regulation of cell adhesion molecules transport to the plasma membrane and actin cytoskeleton reorganization. Regulates the endocytic recycling of occludins, claudins and E-cadherin to the plasma membrane and may thereby regulate the establishment of tight junctions and adherens junctions. In parallel, may regulate actin cytoskeleton reorganization directly through interaction with F-actin or indirectly through actinins and filamins. Most probably involved in the processes of epithelial cell differentiation, cell spreading and neurite outgrowth. [UniProt].
Specifications
| MICALL2 | |
| Polyclonal | |
| Unconjugated | |
| MICALL2 | |
| JRAB; Junctional Rab13-binding protein; MICAL like 2; MICALL2; MICAL-L2; MICAL-like 2; MICAL-like protein 2; milk2; Molecule interacting with CasL-like 2 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 79778 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q8IY33 | |
| MICALL2 | |
| KLH conjugated synthetic peptide between 841-870 amino acids from the C-terminal region of human MILK2. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction