missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICB Antibody (MM0473-3C37), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-11798CL1
This item is not returnable.
View return policy
Description
MICB Monoclonal antibody specifically detects MICB in Human samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| MICB | |
| Monoclonal | |
| CoraFluor 1 | |
| MHC class I chain-related protein B, MHC class I mic-B antigen, MHC class I polypeptide-related sequence B, MIC-B, PERB11.2MHC class I-like molecule PERB11.2-IMX, stress inducible class I homolog | |
| Human recombinant MICB | |
| 0.1 mL | |
| Biologically Active Proteins | |
| 4277 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2 |
| Flow Cytometry, Western Blot, CyTOF | |
| MM0473-3C37 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction