missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICB Monoclonal Antibody (OTI4C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies CF813587
This item is not returnable.
View return policy
Description
MICB Monoclonal antibody specifically detects MICB in Human, Mouse samples. It is validated for Flow CytometrySpecifications
| MICB | |
| Monoclonal | |
| See Label | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P11137, P20357 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 17756, 4133 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry | |
| OTI4C8 | |
| Unconjugated | |
| Map2 | |
| MHC class I chain-related protein B; MHC class I mic-B antigen; MHC class I polypeptide-related sequence B; MHC class I-like located near the LRC, 2; MHC class I-like molecule PERB11.2-IMX; MHC I like leukocyte 2; MICB; MIC-B; Mill2; Mill2 gene for MHC class I-like located near the LRC, 2, exon 3, partial cds, strain:LEW.1lm1; PERB11.2; sMICB; soluble MICB; stress inducible class I homolog | |
| Human recombinant protein fragment corresponding to amino acids 1507-1827 of human MAP2 (NP_002365) produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Lyophilized |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction