Learn More
MICB Monoclonal Antibody (OTI4C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813587
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
MICB Monoclonal antibody specifically detects MICB in Human samples. It is validated for Flow CytometrySpecifications
| MICB | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q29980 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4277 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry | |
| OTI4C8 | |
| Unconjugated | |
| MICB | |
| MHC class I chain-related protein B; MHC class I mic-B antigen; MHC class I polypeptide-related sequence B; MHC class I-like located near the LRC, 2; MHC class I-like molecule PERB11.2-IMX; MHC I like leukocyte 2; MICB; MIC-B; Mill2; Mill2 gene for MHC class I-like located near the LRC, 2, exon 3, partial cds, strain:LEW.1lm1; PERB11.2; sMICB; soluble MICB; stress inducible class I homolog | |
| Human recombinant protein fragment corresponding to amino acids 23-309 of human MICB (NP_005922) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.