Learn More
Micheliolide, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | Micheliolide |
| CAS | 68370-47-8 |
| Molecular Formula | C32 H40 O6 |
| Quantity | 100 mg |
| Synonym | [3aS-(3aalpha,9alpha,9aalpha,9bbeta)]-3a,4,5,7,8,9,9a,9b-Octahydro-9-hydroxy-6,9-dimethyl-3-methylene-azuleno[4,5-b]furan-2(3H)-one |
| InChI Formula | InChI=1S/C32H40O6/c1-2-3-4-5-9-17-32(35-19-20-36-32)18-16-26-27-21-30(33)37-29(27)22-28(26)38-31(34)25-14-12-24(13-15-25)23-10-7-6-8-11-23/h6-8,10-15,26-29H,2-5,9,16-22H2,1H3/t26-,27+,28+,29+/m1/s1 |
| SMILES | CCCCCCCC1(CC[C@H]2[C@H](C[C@@H]3OC(=O)C[C@@H]23)OC(=O)c4ccc(cc4)c5ccccc5)OCCO1 |
| IUPAC Name | [(3aS,4R,5S,6aS)-4-[2-(2-heptyl-1,3-dioxolan-2-yl)ethyl]-2-oxo-3,3a,4,5,6,6a-hexahydrocyclopenta[b]furan-5-yl] 4-phenylbenzoate |
| Molecular Weight (g/mol) | 520.656 |
| Formula Weight | 520.282 |