missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MICL/CLEC12A Antibody (812303) [Janelia Fluor™ 585], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB2950JF585
This item is not returnable.
View return policy
Description
MICL/CLEC12A Monoclonal antibody specifically detects MICL/CLEC12A in Mouse samples. It is validated for Flow Cytometry,CyTOF
Specifications
| MICL/CLEC12A | |
| Monoclonal | |
| Janelia Fluor 585 | |
| CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL | |
| Mouse myeloma cell line NS0-derived recombinant mouse MICL/CLEC12A with an N-terminal 9-His tag, Thr67-Arg267, Accession # Q504P2 | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 812303 | |
| 50mM Sodium Borate | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 160364 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction