Learn More
B Braun™ Straight Tip Jeweler Pattern Micro Forceps
Click to view available options
Length (Metric):
110 mm
Unit Size:
Each
Description
- Stainless steel construction
- Straight tip design
- Manufactured to surgical instrument standards
Specifications
Specifications
| Tip Style | Straight |
| No. per Pack | 1 Pack |
| Material | Stainless Steel |
| Length (Metric) | 110 mm |
| Product Type | Forceps, straight, jeweler, pattern, micro |
| Width (Metric) Tip | 0.15 mm |