missing translation for 'onlineSavingsMsg'
Learn More

Cadence Science™ Micro-Mate™ Interchangeable Syringes

Product Code. 15163265
Change view
Click to view available options
Volume (Metric):
30 mL
Graduations:
1 mL
Connection Type:
Male Luer Lock
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Graduations Connection Type unitSize
15163265 30 mL 1 mL Male Luer Lock Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15163265 Supplier Cadence Science™ Supplier No. 5047

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Syringe barrels and plungers fit perfectly, eliminating the task of matching up syringe parts

Interchangeable syringes are manufactured to ensure the perfect mating of barrels and plungers. Any Micro-Mate interchangeable syringe of a particular size will fit any Micro-Mate interchangeable barrel of that same size.

  • Metal Male Luer Lock, Metal Male Luer or Glass Male Luer tips
  • Constructed of borosilicate glass for heat and corrosion resistance
  • Permanently fused graduations Reinforced flanges with two flat sides prevent syringes from rolling

Specifications

Material Borosilicate Glass
Product Type Interchangeable Syringe
Graduations 1 mL
Volume (Metric) 30 mL
For Use With (Equipment) Micromate Interchangeable Barrel of Same Size
Connection Type Male Luer Lock
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.