Learn More
B Braun™ DUROGRIP™ Micro Needle Holders with Flat Handle
Product Code. 10557860
Change view
Click to view available options
Length (Metric):
145 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
10557860 145 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10557860 Supplier B Braun™ Supplier No. BM002R
  • ONLINE EXCLUSIVE PRICE
    € 441.00
    € 406.10 / Pack of 1
    Save € 34.90
    7% Off
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Safely holds a needle with 0.2mm micro serration. B Braun™ Durogrip™ Micro Needle Holders with Flat Handles include jaws with tungsten carbide inserts.
  • Jaws include tungsten carbide inserts
  • 0.2mm micro serration leads to a safe hold on the needle.
Specifications
Product Type Micro Needle Holder
Material Tungsten Carbide inserts
For Use With (Application) Microsurgery, Cardiovascular surgery
Includes Flat handle
Quantity 1 Pc
Length (Metric) 145 mm