Learn More
Schneider Gerd™ Micro Spatula
Click to view available options
Length (Metric):
130 mm
150 mm
185 mm
210 mm
Unit Size:
Each
Description
- Spatulas are ideal for working with powders, stirring and mixing
- Perfect for miniscule amounts of chemicals
Specifications
Specifications
| Quantity | 1 Pk. |
| Product Type | Micro Spatula |
| Width (Metric) Blade | 5 mm |
| Dimensions (L x W) | 150 x 5 mm |
| Length (Metric) | 150 mm |