Learn More
Sampling Systems™ Micro-Spatulas
Product Code. p-7146963 Shop All Sampling Systems Products
Click to view available options
Length (Metric):
150 mm
210 mm
Length (Metric) Blade:
35 mm
50 mm
Unit Size:
Each
Product Code. 15829581
Brand: Sampling Systems™ A1003
Only null left
Add to basket
Add to basket
These Micro-Spatulas are ideal for handling small quantities of powders and granules.
  • Stainless steel construction
  • Two sizes available
  • Crevice-free
  • Bright polish finish
Specifications
Product Type Micro Spatula
Finish Bright Polish
Length (Metric) Blade 35 mm
Width (Metric) Blade 2.5 mm
Material Stainless Steel
Length (Metric) 150 mm