Learn More
Fisherbrand™ Microcentrifuge Tube, 1.5mL
Product Code. 12942230
Click to view available options
Color:
Clear
Unit Size:
Pack of 1000
Product Code. 12942230
Brand: Fisherbrand™ 200400SP
Only null left
Add to basket
Add to basket
Made from selected high-quality and biologically inert polypropylene. Leak-free caps stay closed during centrifugation with excellent visibility.
  • Polypropylene graduated microcentrifuge tube 
  • Snap top cap
  • With write on label 
Specifications
Cap Style Snap Cap
Closure Color Clear
Closure Material Polypropylene
Capacity (Metric) 1.5 mL
Color Clear
Material Polypropylene
Graduated Yes