missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Microglass Fiber Filters, Grade 264
Suitable for micro-filtration and can be used in various fields.
Brand: Fisherbrand™
Additional Details : Weight : 0.01000kg
Description
- High level of particle retension
- Large load capacity
- High chemical purity (made with demineralized water)
- Non-hygroscopic and biologically inert
- High permeability to air
- Chemical resistance to most solvents and reagents (except hydrofluoric acid)
- Resist to temperature of about 500°C
Specifications
65 g/m2 | |
Plain Disc | |
1.5 μm | |
Air pollution control, suspended solids in the water (in accordance with EN872 standard) by liquid scintillation counting, collection of cells | |
Non-hygroscopic | |
0.011 in. | |
Filter |
Micro filtration | |
Borosilicate Glass | |
60 sec. | |
264 | |
500°C | |
0.28 mm | |
4.92 in. |