Learn More
Hettich Lab Technology™ Microlitre Centrifuge
Description
- Better results and more samples in less time - increase efficiency and throughput in your lab
- Wide range of capabilities due to the variety of models and accessories available
- Pre-cooling function for fast operation - reliable and steady temperature control systems with quick pre-cool and standby functions for precise temperature control
Specifications
Specifications
| Capacity | 30 x 1.5/2 mL |
| Max. Speed | 15,000 rpm |
| Max. RCF | 21,382 x g |
| Safety Features | Lid dropping protection |
| Depth (Metric) | 547 mm |
| Refrigerated | Yes |
| Product Line | MIKRO 200R |
| Product Type | Microlitre Centrifuge |
| Temperature Range (Metric) | -10°C to +40°C |