Learn More
Greiner Bio-One MICROLON™ 96-well ELISA Single-Break Strip Plates
Click to view available options
Color:
Clear
Clear, Blue
Binding Property:
High Binding
Medium Binding
Unit Size:
Pack of 100
Description
- Free of detectable DNase, RNase, human DNA Non-pyrogenic
- Individual wells can be pipetted without leaving residue
- Flat bottom enables precise optical measurements
- Suitable for transmission measurements
- Specifica
Specifications
Specifications
| Breakapart | Single break |
| Binding Property | High Binding |
| Format | Strip Plate |
| Array | 8 x 12 |
| Binding Type | High Binding |
| Color | Clear, Blue |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Well Shape | Curved Edges, Flat Bottom (C-well) |
| Show More |