Learn More
Epredia™ Micromesh™ Biopsy Cassettes in Tube Packs, lilac
Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
Unit Size:
Each
Pack of 10
Description
- Acetal polymer construction
- Large compartment measuring 24 × 27mm is perfect even for needle biopsies
- Recessed lid is specially designed to lie flush with top surface of cassette, which allows cassettes to be labeled on the Shandon™ MicroWriter™ series automatic labelers with the lids on
- Cassette writing face is at a 45° angle
- Cases of 10 tubes of 75 cassettes each with lids attached
Specifications
Specifications
| Pore Style | Small Square Pore |
| Writing Face Angle | 45° |
| Color | Gray |
| Length (Metric) | 24 mm |
| Width (Metric) | 27 mm |
| Material | Acetal Polymer |
| Product Type | Biopsy Cassette |