Learn More
Micronova™ MegaTape™ Polyethylene Cleanroom Tape
Shop All Micronova Products
Click to view available options
Width (Metric):
25.4 mm
50.8 mm
Unit Size:
Each
Descripción
The Micronova MegaTape™ Polyethylene Cleanroom Tape, with hi-tack acrylic adhesive, provides superior sealing abilities and removes cleanly with no adhesive residue. This tape is low outgassing, has good conformability, similar to vinyl tapes, and shows excellent resistance to UV, moisture, acids, alkalis, oils, abrasion and weathering.
Especificaciones
Especificaciones
| Autoclavable | Not Autoclavable |
| Adhesive Type | Acrylic |
| Color | White |
| Material | Polyethylene |
| Product Type | Cleanroom Tape |
| Sterility | Non-sterile |
| For Use With (Application) | Ideal for Clean Construction and Maintenance Projects |
| Temperature Range (English) | 60°F to 80°F |
| Temperature Range (Metric) | 16°C to 27°C |
| Dimensions (LxW) | 33 m x 50.8 mm (36 yd. x 2 in.) |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido