Learn More
Kern Sohn Microscope Camera ODC
Click to view available options
Port Type:
USB 2.0
USB 3.0
Resolution:
20 MP
5.1 MP
Size:
1 in. Sensor
1/2.5 in. Sensor
Compact, 1 in. Sensor
Unit Size:
Each
Description
- The universal microscope cameras can be used anywhere and can be connected to the microscope as well as to a laptop or PC using the USB cable (USB 2.0)
- Through the proven CMOS technology, in connection with the USB 2.0 the images are shown quickly and clearly
- These cameras are also ideal for more demanding applications, such as, for example, darkfield, phase contrast and for fluorescence applications.
- Your daily work is made significantly easier with the very best synchronisation, a high frame rate as well as stable image performance together with our Microscope VIS software which we deliver with the product.
- The power supply is through the USB cable, which means that no additional power supply is required
- As well as the camera, the delivery includes our multi-lingual KERN Microscope VIS Basic KERN OXM 901, a USB cable (Length: 2000 mm) various eyepiece adapters and an object micrometer to calibrate the software.
- Please order the appropriate C-mount adapter to fit your KERN microscope now.
Specifications
Specifications
| For Use With (Equipment) | Microscopes |
| Port Type | USB 2.0 |
| Resolution | 20 MP |
| Sensor Types | CMOS |
| Size | 1 in. Sensor |