Learn More
DuraLab Microscope Eyepiece WF20x/9 mm for DF-100 & DR Series
Product Code. 30280298
Click to view available options
Type:
Widefield Eyepiece
Unit Size:
Each
Product Code. 30280298
Brand: DuraLab DRL0062
Only null left
Add to basket
Add to basket
Designed to achieve high magnification, supporting detailed specimen observation in laboratories and research environments.
  • Provides up to 2000x magnification when paired with a 100x objective lens
  • Enables students to explore detailed cellular structures, including organelles in plant and animal cells
  • Supports viewing of sub-cellular features like mitochondria, chloroplasts, and nuclear details
  • Facilitates study of microorganisms such as bacteria, viruses, and protozoa for enhanced microbiology learning
Specifications
Type Widefield Eyepiece
Compatible with DRL0039 - DuraLab DF-100 Classic Monocular Microscope, DRL0040 - DuraLab DR-410 Advanced Achromatic Monocular Microscope, DRL0041 - DuraLab DR-420 Advanced Achromatic Binocular Microscope, DRL0042 - DuraLab DR-510 Advanced Semi-Plan Monocular Microscope, DRL0043 - DuraLab DR-520 Advanced Semi-Plan Binocular Microscope, DRL0044 - DuraLab DR-530 Advanced Semi-Plan Trinocular Microscope