Learn More
Hirschmann™ Microscope Slides with Cut Edges
Click to view available options
:
Dimensions (L x W): 76 x 26 mm
Description
- Cut Edges
- With LOT number on packaging
- CE acc. to IVD 98/79 EC
Specifications
Specifications
| Length (Metric) | 76 mm |
| Width (Metric) | 26 mm |
| No. per Pack | 100 x 50/Pk. |
| Edges | Cut |
| Thickness (Metric) | 0.95 to 1.05 mm |
| Type | Microscope Slide |
| Color | Extra White |
| Certifications/Compliance | CE acc. IVD 98/79 EC |
| Description | Tropical climate packaging, best before 6 months after delivery |
| Dimensions (L x W) | 76 x 26 mm |