Learn More
Thermo Scientific™ Microscope Slides, Cut, Square
Premium slides are manufactured through a unique proprietary cleaning process intended to provide consistent results and ease interpretation. This process allows these slides to to be used in routine histology and microbiology applications; and specifically in applications by delivering consistently crisp, featherededge blood smears with good morphology.
Merk:  Thermo Scientific™ AAAC0680068002EB0MNZ#0#G#
Overige gegevens : Gewicht : 0.80000kg
 Bekijk meerdere types van dit product via de
Produktcode. 15353786
  • € 369.81 / 100 stuks
Op voorraad 4
Toevoegen aan winkelmandje
Beschrijving
Beschrijving
  • Manufactured from Swiss extra-white soda-lime glass
  • Reduced iron content for low self-fluorescence
  • Greenish shimmer minimized
  • Guaranteed optimum, faultless transparency
  • Clean, flat and suitable for immediate use
Specificatie
Specificatie
68mm
1mm
Uncoated
100 pieces in paper
68 x 68mm
68mm
Extra White
Soda-lime glass
No
Video's