Learn More
Hirschmann™ Microscopic Slide, Cut Edges with Double Frosted End
Examine objects with steadiness. Hirschmann™ Microscopic slides are perfect for sturdy examination of objects under a microscope.
Brand:  Hirschmann™ 8209101
Additional Details : Weight : 0.25000kg
Product Code. 10039700
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Double frosted end
  • Cut Edges
  • With LOT number on packaging
  • CE acc. to IVD 98/79 EC
Specifications
Specifications
Cut
0.95 to 1.05 mm
26 mm
CE, IVD 98/79 EC
76 x 26 mm
Double Frosted
76 mm
White
Extra White