Learn More
Trajan Scientific and Medical™ MicroVolume Syringes
Click to view available options
Volume (Metric):
5 μL
100 μL
Needle Gauge:
25 G
26 G
Diameter (Metric) Outer Needle:
0.47 mm
0.5 mm
Unit Size:
Each
Description
- High quality solutions for liquid handling
- Accurate and reproducible injections
- Manual injection syringe
Specifications
Specifications
| Description | Guided Plunger Syringe |
| Needle Gauge | 26 G |
| Diameter (Metric) Inner Needle | 0.11 mm |
| Diameter (Metric) Outer Needle | 0.47 mm |
| Product Type | Removable Needle |
| Volume (Metric) | 5 μL |
| Needle Point Style | Bevel |