missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MID1 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-26612AF750
This item is not returnable.
View return policy
Description
MID1 Polyclonal antibody specifically detects MID1 in Human samples. It is validated for Western Blot
Specifications
| MID1 | |
| Polyclonal | |
| Western Blot | |
| EC 6.3.2.-, FLJ57031, FLJ58683, FLJ76288, FXYGBBB1, Midin, midline 1 (Opitz/BBB syndrome), RNF59MIDIN, TRIM18mouse, homolog of | |
| The immunogen recognized by this antibody maps to a region between residue 617 and 667 of human midline 1 using the numbering given in entry NP_000372.1 (GeneID 4281). | |
| 0.1 mL | |
| Lipid and Metabolism, Zinc Finger | |
| 4281 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction