missing translation for 'onlineSavingsMsg'
Learn More
Learn More
midA Antibody (OTI4A5), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72681FR
This item is not returnable.
View return policy
Description
midA Monoclonal specifically detects midA in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| midA | |
| Monoclonal | |
| DyLight 680 | |
| PBS with 0.05% Sodium Azide | |
| NDUFAF7 | |
| Full length recombinant protein of human midA (NP_653337) produced in HEK293T cells. | |
| 0.1 mL | |
| 55471.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI4A5 | |
| Western Blot, Immunohistochemistry | |
| chromosome 2 open reading frame 56, MidA, mitochondrial, PRO1853 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction