missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Midkine Antibody (OTI6C8), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72682H
This item is not returnable.
View return policy
Description
Midkine Monoclonal specifically detects Midkine in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Midkine | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| MDK | |
| Recombinant protein fragment corresponding to amino acids 21-143 of human Midkine (NP_001012334) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot | |
| OTI6C8 | |
| Western Blot | |
| Amphiregulin-associated protein, Midgestation and kidney protein, midkine, midkine (neurite growth-promoting factor 2), MK1, MKARAP, NEGF2FLJ27379, Neurite outgrowth-promoting factor 2, Neurite outgrowth-promoting protein | |
| Mouse | |
| Affinity Purified | |
| Biologically Active Proteins, Cell Cycle and Replication | |
| 4192.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction